Learn More
uvex Phynomic AirLite A ESD Safety Glove
The Uvex phynomic airLite ESD is a light safety glove that offers a noticeable difference in wearer comfort.
Brand:  uvex 6003810
 View more versions of this product
Product Code. 17717579
  • 463.00 DKK / 10 pairs
Only null left
Estimated Shipment: 31-07-2025
Add to basket
This item is not returnable. View return policy
Description
Description
  • The lightest and most sensitive uvex phynomic cut protection glove in Cut Level A
  • Outer material made from polyamide, elastane and carbon
  • Breathable 'airLite aqua-polymer coating on palm and fingertips
  • Safety glove with touchscreen function – for use on almost all screens, tablets and mobile phones
  • Safety glove with ESD function in line with DIN EN 16350:2014 – protects workpiece against electrostatic charge
  • Good grip in dry and slightly damp areas
  • Free from accelerators, and health protection and skin compatibility dermatologically approved (proDERM)
  • Very lightweight and breathable safety glove
  • Outstanding sensitivity thanks to new breathable 'airLite' aqua-polymer coating combined with fine liner (18 gauge)
  • Safety gloves with knitted cuff and ergonomic fit
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EN 388 (3 1 1 0 X) and DIN EN 16350:2014
Black
Smooth
Not tested
Polyamide, Elastane, Carbon Fiber
ESD Gloves
Non-sterile
Aqua-Polymer Foam
Reusable
Ambidextrous
Yes
Powder Free
10
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
uvex Phynomic AirLite A ESD Safety Glove > Size: 10