Få mere at vide
Phthivazid, LoGiCal
Artikelnummer. 30463219
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30463219 10 mg
1 options
Artikelnummer. 30463219 Leverandør LoGiCal Leverandørnr. LGCFOR3191.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phthivazid
CAS 149-17-7
Analyt- eller komponentnavne Phthivazid
Forsendelsestilstand Room Temperature
Molekylær formel C14 H13 N3 O3
Certificeringer/compliance ISO 9001
Synonym 4-Pyridinecarboxylic acid, 2-[(4-hydroxy-3-methoxyphenyl)methylene]hydrazide, 4-Pyridinecarboxylic acid, [(4-hydroxy-3-methoxyphenyl)methylene]hydrazide (9CI), Isonicotinic acid, vanillylidenehydrazide (6CI,8CI), Ftivazid, Ftivazide, N-Isonicotinamido-3-methoxy-4-hydroxybenzalimine, NSC 31725, Phthivazid, Phthivazide, Vanicid, Vanillaberon, Vanizide
InChI formel InChI=1S/C14H13N3O3/c1-20-13-8-10(2-3-12(13)18)9-16-17-14(19)11-4-6-15-7-5-11/h2-9,18H,1H3,(H,17,19)
Løsningstype Neat
SMIL COc1cc(C=NNC(=O)c2ccncc2)ccc1O
IUPAC navn N-[(4-hydroxy-3-methoxyphenyl)methylideneamino]pyridine-4-carboxamide
Industry Type Clinical Testing
Molekylvægt (g/mol) 271.27
Mængde 10 mg
Formel vægt 271.0957 g/mol