missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Phthaloyl Amlodipine, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 3-Ethyl 5-Methyl (4RS)-4-(2-Chlorophenyl)-2-([2-(1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)ethoxy]methyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate |
| CAS | 88150-62-3 |
| Renhedsgrad noter | HPLC |
| Molekylær formel | C28 H27 Cl N2 O7 |
| Mængde | 250 mg |
| Synonym | 3-Ethyl 5-Methyl (4RS)-4-(2-Chlorophenyl)-2-[[2-(1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)ethoxy]methyl]-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate, Amlodipine Besilate Imp. A (EP), Phthaloyl Amlodipine |
| InChI formel | InChI=1S/C28H27ClN2O7/c1-4-38-28(35)24-21(15-37-14-13-31-25(32)17-9-5-6-10-18(17)26(31)33)30-16(2)22(27(34)36-3)23(24)19-11-7-8-12-20(19)29/h5-12,23,30H,4,13-15H2,1-3H3 |
| SMIL | CCOC(=O)C1=C(COCCN2C(=O)c3ccccc3C2=O)NC(=C(C1c4ccccc4Cl)C(=O)OC)C |
| IUPAC navn | 3-O-ethyl 5-O-methyl 4-(2-chlorophenyl)-2-[2-(1,3-dioxoisoindol-2-yl)ethoxymethyl]-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate |
| Molekylvægt (g/mol) | 538.98 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?