Få mere at vide
Phthalimide-d4, TRC
Artikelnummer. 30422685
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30422685 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30422685 Leverandør Toronto Research Chemicals Leverandørnr. TRCP38455210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phthalimide-d4
CAS 60161-31-1
Molekylær formel C8HD4NO2
Mængde 10 mg
Synonym 1H-Isoindole-1,3(2H)-dione-4,5,6,7-d4, 1,2-Benzenedicarboximide-d4, 1,3-Dihydro-2H-isoindole-1,3-dione-d4, 1,3-Dioxo-1,3-dihydroisoindole-d4, 1,3-Isoindolinedione-d4, Benzoimide-d4, Isoindole-1,3-dione-d4, Kladnoite-d4, Levegal PEW-T-d4, NSC 3108-d4, Phenylimide-d4, Phthalic Dicarboximide-d4
InChI formel InChI=1S/C8H5NO2/c10-7-5-3-1-2-4-6(5)8(11)9-7/h1-4H,(H,9,10,11)/i1D,2D,3D,4D
SMIL Cl.NCCc1c(OC)cc(CC)c(OC)c1
IUPAC navn 4,5,6,7-tetradeuterioisoindole-1,3-dione
Molekylvægt (g/mol) 151.16
Formel vægt 151.0571