missing translation for 'onlineSavingsMsg'
Få mere at vide

Phthalic Acid Di(methyl-d3) Ester, TRC

Artikelnummer. 30372935
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30372935 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30372935 Leverandør Toronto Research Chemicals Leverandørnr. TRCD47797610MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Phthalic Acid Di(methyl-d3) Ester
CAS 85448-30-2
Molekylær formel C10 D6 H4 O4
Mængde 10 mg
Synonym 1,2-Benzenedicarboxylic Acid Di(methyl-d3) Ester, 1,2-Benzenedicarboxylic acid, di(methyl-d3) ester (9CI), Phthalic acid, di(methyl-d3) ester (7CI), 1,2-Di(methyl-d3) 1,2-benzenedicarboxylate, Phthalic acid dimethyl ester D6, Phthalic acid dimethyl ester-d6, Phthalic acid dimethyl ester D6 (dimethyl-D6), Dimethyl-d6 phthalate, Phthalic acid, bis-methyl ester D6
InChI formel InChI=1S/C10H10O4/c1-13-9(11)7-5-3-4-6-8(7)10(12)14-2/h3-6H,1-2H3/i1D3,2D3
SMIL [2H]C([2H])([2H])OC(=O)c1ccccc1C(=O)OC([2H])([2H])[2H]
IUPAC navn bis(trideuteriomethyl) benzene-1,2-dicarboxylate
Molekylvægt (g/mol) 200.22
Formel vægt 200.0956
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.