Få mere at vide
Phthalic Acid-d4, TRC
Artikelnummer. 30411694
Skift visning
Mængde:
2.5 g
Pakningsstørrelse:
2.5g
Artikelnummer. Mængde unitSize
30411694 2.5 g 2.5g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30411694 Leverandør Toronto Research Chemicals Leverandørnr. TRCP3844822.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phthalic Acid-d4
CAS 87976-26-9
Renhedsgrad noter HPLC
Molekylær formel C8 2H4 H2 O4
Mængde 2.5 g
Synonym 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, 3,4,5,6-Tetradeuteriophthalic acid, Benzene-3,4,5,6-d4-1,2-dicarboxylic acid, Phthalic acid-d4, Phthalic-d4 acid, Phthalic-3,4,5,6-D4 acid
InChI formel InChI=1S/C8H6O4/c9-7(10)5-3-1-2-4-6(5)8(11)12/h1-4H,(H,9,10)(H,11,12)/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c([2H])c(C(=O)O)c(C(=O)O)c1[2H]
IUPAC navn 3,4,5,6-tetradeuteriophthalic acid
Molekylvægt (g/mol) 170.16
Formel vægt 170.0517
Procent renhed >95