Få mere at vide
Phthalic acid, bis-methyl ester D4 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Artikelnummer. 30313026
Skift visning
Mængde:
1 mL
Artikelnummer. Mængde
30313026 1 mL
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313026 Leverandør Dr. Ehrenstorfer Leverandørnr. DREA16174010AL100
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phthalic acid, bis-methyl ester D4
CAS 93951-89-4
Opløsningsmiddel eller Matrix Acetonitrile
Molekylær formel C10 2H4 H6 O4
Mængde 1 mL
Synonym 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, 1,2-dimethyl ester, 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, dimethyl ester (9CI), Phthalic acid dimethyl ester D4
InChI formel InChI=1S/C10H10O4/c1-13-9(11)7-5-3-4-6-8(7)10(12)14-2/h3-6H,1-2H3/i3D,4D,5D,6D
SMIL [2H]c1c([2H])c([2H])c(C(=O)OC)c(C(=O)OC)c1[2H]
IUPAC navn dimethyl 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate
Molekylvægt (g/mol) 198.21
Formel vægt 198.083 g/mol