Få mere at vide
Phthalic acid, bis-ethyl ester D4 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Artikelnummer. 30327932
Skift visning
Mængde:
1 mL
Artikelnummer. Mængde
30327932 1 mL
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30327932 Leverandør Dr. Ehrenstorfer Leverandørnr. DREA16172010AL100
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phthalic acid, bis-ethyl ester D4
CAS 93952-12-6
Opløsningsmiddel eller Matrix Acetonitrile
Molekylær formel C12 2H4 H10 O4
Mængde 1 mL
Synonym 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, diethyl ester (9CI), Phthalic acid diethyl ester (3,4,5,6)-D4, Phthalic acid, bis-ethyl ester D4, Diethyl phthalate-d4
InChI formel InChI=1S/C12H14O4/c1-3-15-11(13)9-7-5-6-8-10(9)12(14)16-4-2/h5-8H,3-4H2,1-2H3/i5D,6D,7D,8D
SMIL [2H]c1c([2H])c([2H])c(C(=O)OCC)c(C(=O)OCC)c1[2H]
IUPAC navn diethyl 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate
Molekylvægt (g/mol) 226.26
Formel vægt 226.1143 g/mol