Få mere at vide
Phthalic acid, bis-butyl ester D4 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Artikelnummer. 30325874
Skift visning
Mængde:
1 mL
Artikelnummer. Mængde
30325874 1 mL
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325874 Leverandør Dr. Ehrenstorfer Leverandørnr. DREA16171010AL100
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phthalic acid, bis-butyl ester D4
CAS 93952-11-5
Opløsningsmiddel eller Matrix Acetonitrile
Molekylær formel C16 2H4 H18 O4
Mængde 1 mL
InChI formel InChI=1S/C16H22O4/c1-3-5-11-19-15(17)13-9-7-8-10-14(13)16(18)20-12-6-4-2/h7-10H,3-6,11-12H2,1-2H3/i7D,8D,9D,10D
SMIL [2H]c1c([2H])c([2H])c(C(=O)OCCCC)c(C(=O)OCCCC)c1[2H]
IUPAC navn dibutyl 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate
Molekylvægt (g/mol) 282.37
Formel vægt 282.1769 g/mol