Få mere at vide
Phthalic Acid Bis(3,7-dimethyloctyl) Ester-d4, TRC
Artikelnummer. 30179750
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179750 10 mg
30180373 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30179750 Leverandør Toronto Research Chemicals Leverandørnr. TRCP38453710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1398065-81-0
Molekylær formel C28 D4 H42 O4
Molekylvægt (g/mol) 450.6871
Synonym 1,2-Benzenedicarboxylic-3,4,5,6-d4 acid bis(3,7-dimethyloctyl) ester, 1,2-Bis(3,7-dimethyloctyl) 1,2-benzenedicarboxylate-3,4,5,6-d4, Phthalic-d4 acid bis(3,7-dimethyloctyl) ester, Bis(3,7-dimethyloctyl) phthalate-3,4,5,6-d4, Phthalic Acid Bis(3,7-dimethyloctyl) Ester-D4 (phthalic-D4)
IUPAC navn bis(3,7-dimethyloctyl) 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate
SMIL [2H]c1c([2H])c([2H])c(C(=O)OCCC(C)CCCC(C)C)c(C(=O)OCCC(C)CCCC(C)C)c1[2H]
Tekniske data
Kemisk navn eller materiale Phthalic Acid Bis(3,7-dimethyloctyl) Ester-d4
Mængde 10 mg
InChI formel InChI=1S/C28H46O4/c1-21(2)11-9-13-23(5)17-19-31-27(29)25-15-7-8-16-26(25)28(30)32-20-18-24(6)14-10-12-22(3)4/h7-8,15-16,21-24H,9-14,17-20H2,1-6H3/i7D,8D,15D,16D
Formel vægt 450.3647