Få mere at vide
Phthalic Acid Anhydride-d4, TRC
Artikelnummer. 30426720
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30426720 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30426720 Leverandør Toronto Research Chemicals Leverandørnr. TRCP384488100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phthalic Acid Anhydride-d4
CAS 75935-32-9
Molekylær formel C8 D4 O3
Mængde 100 mg
Synonym 1,3-Isobenzofurandione-4,5,6,7-d4 (9CI','ACI), Phthalic-d4 anhydride (7CI), Phthalic Anhydride-D4, Phthalic Acid Anhydride-d4
InChI formel InChI=1S/C8H4O3/c9-7-5-3-1-2-4-6(5)8(10)11-7/h1-4H/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c([2H])c2C(=O)OC(=O)c2c1[2H]
IUPAC navn 4,5,6,7-tetradeuterio-2-benzofuran-1,3-dione
Molekylvægt (g/mol) 152.14
Formel vægt 152.041