Få mere at vide
Phthalic-3,4,5,6-d4 Acid, CDN
Artikelnummer. 30321156
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30321156 5 g
30313084 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321156 Leverandør CDN Leverandørnr. CDND23765G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phthalic Acid-d4
CAS 87976-26-9
Molekylær formel C8 2H4 H2 O4
Mængde 5 g
InChI formel InChI=1 S/C8H6O4/c9-7(10)5-3-1-2-4-6(5)8(11)12/h1-4 H,(H,9,10)(H,11,12)/i1D,2 D,3 D,4 D
SMIL [2 H]c1c([2 H])c([2 H])c(C(=O)O)c(C(=O)O)c1[2 H]
IUPAC navn 3,4,5,6-tetradeuteriophthalic acid
Molekylvægt (g/mol) 170.16
Formel vægt 170.0517 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity