missing translation for 'onlineSavingsMsg'
Learn More

Phthalhydrazide, 98%, Thermo Scientific Chemicals

Catalog Number p-7022033
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
1445-69-8
C8H6N2O2
162.15
MFCD00006888
JNQUJPBFSQETPT-UHFFFAOYNA-N
2,3-dihydro-1,4-phthalazinedione, phthalazine-1,4-diol, phtalylhydrazine, 1,2,3,4-tetrahydrophthalazine-1,4-dione, 1,4-phthalazinedione, 2,3-dihydro, 1 2h-phthalazinone, 4-hydroxy, 2,3-dihydro-phthalazine-1,4-dione, phthalhydrazine, phthalylhydrazine, phthaloylhydrazine
219401
O=C1N=NC(=O)C2C=CC=CC12
This item is not returnable. View return policy
1445-69-8
C8H6N2O2
162.15
MFCD00006888
JNQUJPBFSQETPT-UHFFFAOYNA-N
2,3-dihydro-1,4-phthalazinedione, phthalazine-1,4-diol, phtalylhydrazine, 1,2,3,4-tetrahydrophthalazine-1,4-dione, 1,4-phthalazinedione, 2,3-dihydro, 1 2h-phthalazinone, 4-hydroxy, 2,3-dihydro-phthalazine-1,4-dione, phthalhydrazine, phthalylhydrazine, phthaloylhydrazine
219401
O=C1N=NC(=O)C2C=CC=CC12

Phthalhydrazide is a reagent used in the synthesis of various pyrazolophthalazine derivatives is and used in medicine double hydrazine phthalocyanine work intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phthalhydrazide is a reagent used in the synthesis of various pyrazolophthalazine derivatives is and used in medicine double hydrazine phthalocyanine work intermediates.

Solubility
Soluble in acetone and acetic acid.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 342°C to 345°C
Quantity 10 g
Beilstein 135926
IUPAC Name 1,4,4a,8a-tetrahydrophthalazine-1,4-dione
Formula Weight 162.15
Percent Purity 98%
Chemical Name or Material Phthalhydrazide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'