Learn More
Phthalazine, 98%, BLD Pharm™
Product Code. 30940015
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
Unit Size:
100g
10g
1g
25g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30940015 5 g 5g
30929474 1 g 1g
30935157 25 g 25g
30936781 100 g 100g
30938225 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 30940015 Supplier BLD Pharm Supplier No. BD1058875g
 Restricted Item
Forventet forsendelse: 27-11-2026
Specifications
CAS 253-52-1
Molecular Formula C8H6N2
MDL Number MFCD00006908
Quantity 5 g
SMILES C12=C(C=NN=C2)C=CC=C1
Molecular Weight (g/mol) 130.15
Percent Purity 98%
Chemical Name or Material Phthalazine