missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus(V) oxychloride, 99%, Thermo Scientific Chemicals

Catalog Number 15480987
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
10025-87-3
Cl3OP
153.323
MFCD00011443
XHXFXVLFKHQFAL-UHFFFAOYSA-N
phosphorus oxychloride, phosphoryl chloride, phosphoric trichloride, phosphoryl trichloride, phosphoroxychloride, trichlorophosphine oxide, phosphorus oxytrichloride, fosforoxychlorid, phosphorous oxychloride, phosphorylchlorid
24813
CHEBI:30336
O=P(Cl)(Cl)Cl
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
10025-87-3
Cl3OP
153.323
MFCD00011443
XHXFXVLFKHQFAL-UHFFFAOYSA-N
phosphorus oxychloride, phosphoryl chloride, phosphoric trichloride, phosphoryl trichloride, phosphoroxychloride, trichlorophosphine oxide, phosphorus oxytrichloride, fosforoxychlorid, phosphorous oxychloride, phosphorylchlorid
24813
CHEBI:30336
O=P(Cl)(Cl)Cl

Phosphorus(V) oxychloride is widely used as a dehydrating agent and a chlorinating agent. Useful for the conversion of amide to nitriles, Bischler-Napieralski reaction, Vilsmeier Haak reaction, and other reactions producing commercially important intermediates. Acts a source of liquid phosphorus in semiconductor industry. Manufacture of trialkyl phosphate esters and triarylphosphate esters, including commercially popular tricresyl phosphate,which is useful as a plasticizer, flame retardant, gasoline additive, hydrolytic fluid, etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 105.8°C
Melting Point 1.25°C
Color Colorless
Physical Form Liquid
Quantity 250 g
Assay Percent Range 99%
Linear Formula POCl3
UN Number UN1810
Merck Index 14,7349
Solubility Information Phosphorus(V) oxychloride is soluble in many organic solvents.
Formula Weight 153.33
Percent Purity 99%
Odor Pungent
Refractive Index 1.46
Sensitivity Moisture sensitive
Density 1.645 g/mL
Chemical Name or Material Phosphorus(V) oxychloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phosphorus(V) oxychloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H372-Causes damage to organs.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : phosphorus oxychloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'