Få mere at vide
Phosphoric Acid Tripropyl Ester, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Tripropyl Phosphate |
| CAS | 513-08-6 |
| Molekylær formel | C9 H21 O4 P |
| Mængde | 25 g |
| Synonym | Phosphoric acid','tripropyl ester (8CI','9CI','ACI), Propyl phosphate ((PrO)3PO) (6CI), Tri-n-propyl phosphate, Tripropyl phosphate |
| InChI formel | InChI=1S/C9H21O4P/c1-4-7-11-14(10,12-8-5-2)13-9-6-3/h4-9H2,1-3H3 |
| SMIL | CCCOP(=O)(OCCC)OCCC |
| IUPAC navn | tripropyl phosphate |
| Molekylvægt (g/mol) | 224.23 |
| Formel vægt | 224.1177 |