missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Phosphoric Acid Tripropyl Ester, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Tripropyl Phosphate |
| CAS | 513-08-6 |
| Molekylær formel | C9 H21 O4 P |
| Mængde | 50 g |
| Synonym | Phosphoric acid','tripropyl ester (8CI','9CI','ACI), Propyl phosphate ((PrO)3PO) (6CI), Tri-n-propyl phosphate, Tripropyl phosphate |
| InChI formel | InChI=1S/C9H21O4P/c1-4-7-11-14(10,12-8-5-2)13-9-6-3/h4-9H2,1-3H3 |
| SMIL | CCCOP(=O)(OCCC)OCCC |
| IUPAC navn | tripropyl phosphate |
| Molekylvægt (g/mol) | 224.23 |
| Formel vægt | 224.1177 |
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?