missing translation for 'onlineSavingsMsg'
Få mere at vide

Phosphoric Acid Triheptyl Ester, TRC

Artikelnummer. 30419178
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30419178 1 g
30416152 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30419178 Leverandør Toronto Research Chemicals Leverandørnr. TRCP3594501G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 4621-50-5
Molekylær formel C21H45O4P
Molekylvægt (g/mol) 392.55
Synonym Heptanol Phosphate, Heptyl Phosphate ((C7H15O)3PO) (7CI), Triheptyl Phosphate
IUPAC navn triheptyl phosphate
SMIL CCCCCCCOP(=O)(OCCCCCCC)OCCCCCCC

Tekniske data

Kemisk navn eller materiale Phosphoric Acid Triheptyl Ester
Mængde 1 g
InChI formel InChI=1S/C21H45O4P/c1-4-7-10-13-16-19-23-26(22,24-20-17-14-11-8-5-2)25-21-18-15-12-9-6-3/h4-21H2,1-3H3
Formel vægt 392.3055
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.