Få mere at vide
Phosphoric Acid Dioctyl Ester Potassium Salt, TRC
Artikelnummer. 30423883
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30423883 250 mg
1 options
Artikelnummer. 30423883 Leverandør Toronto Research Chemicals Leverandørnr. TRCP359420250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phosphoric Acid Dioctyl Ester Potassium Salt
CAS 19045-78-4
Molekylær formel C16H34KO4P
Mængde 250 mg
Synonym Dioctyl Phosphate Potassium Salt, Potassium Dioctyl Phosphate
InChI formel InChI=1S/C16H35O4P.K/c1-3-5-7-9-11-13-15-19-21(17,18)20-16-14-12-10-8-6-4-2;/h3-16H2,1-2H3,(H,17,18);/q;+1/p-1
SMIL O=P(OCCCCCCCC)(OCCCCCCCC)[O-].[K+]
IUPAC navn potassium dioctyl phosphate
Molekylvægt (g/mol) 360.51
Formel vægt 360.18