Få mere at vide
Phosphoric Acid-d3 (~85% w/w in D2O), TRC
Artikelnummer. 30422748
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30422748 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30422748 Leverandør Toronto Research Chemicals Leverandørnr. TRCP359202100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phosphoric Acid-d3
CAS 14335-33-2
Molekylær formel 2H3 O4 P
Mængde 100 mg
Synonym Phosphoric acid-d3 (6CI','7CI','8CI','9CI), Deuterium phosphate (D3PO4), Phosphoric acid (D3PO4), Phosphoric acid D3, [2H3]Phosphoric acid
InChI formel InChI=1S/H3O4P/c1-5(2,3)4/h(H3,1,2,3,4)/i/hD3
SMIL [2H]OP(=O)(O[2H])O[2H]
IUPAC navn trideuterio phosphate
Molekylvægt (g/mol) 101.01
Formel vægt 100.9957
Koncentration eller sammensætning (efter analyt eller komponenter) ~85%