missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Phosphoric Acid-d3 (~85% w/w in D2O), TRC
Specifications
Specifications
| Kemisk navn eller materiale | Phosphoric Acid-d3 |
| CAS | 14335-33-2 |
| Molekylær formel | 2H3 O4 P |
| Mængde | 1 g |
| Synonym | Phosphoric acid-d3 (6CI','7CI','8CI','9CI), Deuterium phosphate (D3PO4), Phosphoric acid (D3PO4), Phosphoric acid D3, [2H3]Phosphoric acid |
| InChI formel | InChI=1S/H3O4P/c1-5(2,3)4/h(H3,1,2,3,4)/i/hD3 |
| SMIL | [2H]OP(=O)(O[2H])O[2H] |
| IUPAC navn | trideuterio phosphate |
| Molekylvægt (g/mol) | 101.01 |
| Formel vægt | 100.9957 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?