Få mere at vide
Phosphoric Acid-d3 (85% w/w in D2O), CDN
Artikelnummer. 30323403
Skift visning
Mængde:
100 g
Artikelnummer. Mængde
30323403 100 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323403 Leverandør CDN Leverandørnr. CDND0616100G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phosphoric Acid-d3
CAS 14335-33-2
Opløsningsmiddel eller Matrix D2O
Molekylær formel 2H3 O4 P
Mængde 100 g
InChI formel InChI=1 S/H3O4P/c1-5(2,3)4/h(H3,1,2,3,4)/i/hD3
SMIL [2 H]OP(=O)(O[2 H])O[2 H]
IUPAC navn trideuterio phosphate
Molekylvægt (g/mol) 101.01
Formel vægt 100.9957 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity
Koncentration eller sammensætning (efter analyt eller komponenter) 85 %