Få mere at vide
Phosphoglycolic Acid, TRC
Artikelnummer. 30425310
Skift visning
Mængde:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30425310 5 mg
30413845 25 mg
30424631 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30425310 Leverandør Toronto Research Chemicals Leverandørnr. TRCP3582005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 13147-57-4
Molekylær formel C2H5O6P
Molekylvægt (g/mol) 156.03
Synonym 2-(Phosphonooxy)acetic Acid, (Phosphonooxy)acetic Acid, Glycolic Acid Phosphate, Glycolic Acid, di-H Phosphate, Glycolic Acid, Dihydrogen Phosphate, 2-Phosphoglycolic Acid, Glycophosphoric Acid
IUPAC navn 2-phosphonooxyacetic acid
SMIL OC(=O)COP(=O)(O)O
Tekniske data
Kemisk navn eller materiale Phosphoglycolic Acid
Renhedsgrad noter HPLC
Mængde 5 mg
InChI formel InChI=1S/C2H5O6P/c3-2(4)1-8-9(5,6)7/h1H2,(H,3,4)(H2,5,6,7)
Formel vægt 155.9824
Procent renhed >95%