Få mere at vide
Phosphocholine-d9 Chloride Calcium Salt, TRC
Artikelnummer. 30411276
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30411276 5 mg
30425051 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30411276 Leverandør Toronto Research Chemicals Leverandørnr. TRCP3545025MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 344299-43-0
Molekylær formel C5H4D9CaClNO4P
Molekylvægt (g/mol) 266.72
Synonym N,N,N-Trimethyl-2-(phosphonooxy)ethanaminium-d9 Chloride Calcium Salt, Biocalcose-d9, Calcium Phosphorylcholine-d9, Calcium Phosphorylcholine-d9 Chloride, Colifos-d9, Colincalcium-d9, Epafosforil-d9, Fosfocolina-d9, Ipercolin-d9, Isocolin-d9, Mebophos-d9, Merival-d9, Pancholin-d9, Phoscholin-d9
IUPAC navn calcium 2-(tris(methyl-d3)ammonio)ethyl phosphate chloride
SMIL O=P([O-])([O-])OCC[N+](C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H].[Ca+2].[Cl-]
Tekniske data
Kemisk navn eller materiale Phosphocholine-d9 Chloride Calcium Salt
Mængde 5 mg
InChI formel InChI=1S/C5H14NO4P.Ca.ClH/c1-6(2,3)4-5-10-11(7,8)9;;/h4-5H2,1-3H3,(H-,7,8,9);;1H/q;+2;/p-2/i1D3,2D3,3D3;;
Formel vægt 266.05