missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ Phospho-Paxillin (Tyr118) Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 13207948
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
20 μg
Pakningsstørrelse:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
13207948 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 13207948 Leverandør Invitrogen™ Leverandørnr. PA523327

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Polyclonal Antibody

Sequence homology: Opossum/Chimpanzee/Orangutan/Salmon (100%), Tetradon (85%).

Paxillin is a focal adhesion protein and a substrate for several tyrosine kinases such as src, FAK, and p210BRC/ABL. The tyrosine phosphorylation of paxillin is affected by conditions that change cell-cell adhesion. Paxillin associates tightly with FAK and Crk through its SH2 domain. This interaction is independent of the extracellular matrix. Although paxillin was initially discovered in fibroblasts, its phosphorylation may also be important during neurite extension during differentiation.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen Phospho-Paxillin (Tyr118)
Applikationer Western Blot
Klassifikation Polyclonal
Koncentration 0.2 mg/mL
Konjugeret Unconjugated
Formulering PBS with 0.05% BSA and 0.05% sodium azide
Gene PXN
Genadgangsnr. P49023, P49024, Q66H76, Q8VI36
Gene Alias adaptor protein; AW108311; AW123232; FLJ16691; hCG_1778014; I79_001229; myocardial ischemic preconditioning associated protein 7; Pax; PAX-1; PAXI; Paxillin; paxillin a; paxillin alpha; pxn; pxna; testicular tissue protein Li 134; TGFB1I1; transforming growth factor beta 1 induced transcript 1; wu:fw71f12
Gensymboler PXN, pxna
Værtsarter Rabbit
Immunogen A portion of human Paxillin protein containing a phosphorylated tyrosine residue at position 118.
Oprensningsmetode Antigen affinity chromatography
Mængde 20 μg
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 100353305, 19303, 360820, 395832, 399546, 486299, 5829
Målarter Human, Mouse, Rat, Canine, Rabbit, Monkey, Chicken, Frog, Zebrafish, Rhesus Macaque
Indhold og opbevaring -20°C, Avoid Freeze/Thaw Cycles
Produkttype Antibody
Form Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.