missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ Phospho-p130 (Ser952) Recombinant Rabbit Monoclonal Antibody (K01_1X68)
GREENER_CHOICE

Artikelnummer. 30825640
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30825640 100 μL
30825907 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30825640 Leverandør Invitrogen™ Leverandørnr. MA563604

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Recombinant Monoclonal Antibody

Phospho-p130 (Ser952) Recombinant Monoclonal Antibody for ICC/IF, IHC (P), Western Blot

RBL2 (also known as p130) or Retinoblastoma-like protein 2 is a key regulator of entry into cell division. It is directly involved in heterochromatin formation by maintaining overall chromatin structure and (in particular) that of constitutive heterochromatin by stabilizing histone methylation. RBL2 recruits and targets histone methyltransferases KMT5B and KMT5C leading to epigenetic transcriptional repression. It controls histone H4 'Lys20' trimethylation. It probably also acts as a transcription repressor by recruiting chromatin-modifying enzymes to promoters. RBL2 serves as a potent inhibitor of E2F-mediated trans-activation and associates preferentially with E2F5.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen Phospho-p130 (Ser952)
Applikationer Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Klassifikation Recombinant Monoclonal
Klon K01_1X68
Koncentration 1 mg/mL
Konjugeret Unconjugated
Formulering 50mM tris glycine with 150mM NaCl, 0.05% BSA, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene RBL2
Genadgangsnr. Q08999
Gene Alias 130 kDa retinoblastoma-associated protein; FLJ26459; p130; PPAR-alpha-interacting complex protein 128; PRB2; PRIC128; RB transcriptional corepressor like 2; Rb2; rb2 p130; Rbl2; RBR-2; retinoblastoma-like 2; retinoblastoma-like 2 (p130); retinoblastoma-like protein 2; Retinoblastoma-related protein 2
Gensymboler RBL2
Værtsarter Rabbit
Immunogen A synthetic phosphopeptide corresponding to residues surrounding Ser952 of human Rb2 p130.
Oprensningsmetode Protein A/G
Mængde 100 μL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 5934
Målarter Human
Indhold og opbevaring Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Produkttype Antibody
Form Liquid
Isotype IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.