missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ Phospho-IRS1 (Ser616) Monoclonal Antibody (738710)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561370
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
IRS-1, a major substrate of the insulin receptor, is phosphorylated in response to stimulation of cells by insulin, insulin-like growth factor 1 (IGF-1) and interleukin 4 (IL-4). IRS-1 is phosphorylated on serine, threonine and tyrosine residues in a variety of tissues. An insulin-sensitive serine/threonine kinase casein kinase II mediates a portion of the insulin-stimulated serine/threonine phosphorylation of overexpressed IRS-1 in vivo. Thr 502 is identified as the major casein kinase II-catalyzed phosphorylation site in rat IRS-1, and Ser 99 is an additional phosphorylation site catalyzed by casein kinase II. Thus, casein kinase II-catalyzed phosphorylation of IRS-1 may be a component of the intracellular insulin signaling cascade. IRS-1 contains three putative binding sites for 14-3-3 (Ser 270, Ser 374 and Ser 641) and the motif around Ser 270 is located in the phosphortyrosine binding domain of IRS-1, which is responsible for the interaction with the insulin receptor.
Tekniske data
| Phospho-IRS1 (Ser616) | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P35568 | |
| IRS1 | |
| Phosphopeptide containing the human IRS1 S616 site. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 738710 | |
| Unconjugated | |
| IRS1 | |
| G972R; HIRS-1; Insulin receptor substrate 1; IRS1; IRS-1; IRS1IRM; pp185 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 3667 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion