missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ GSK3 beta (Phospho) [pS9] Multispecies InstantOne™ ELISA Kit
Klik for at se tilgængelige muligheder
Mængde:
96 Tests
Pakningsstørrelse:
96 tests
Beskrivelse
Indeholder
- User Manual
- InstantOne™ Assay Plate - 96 well (sealed)
- Cell Lysis Buffer (5X)
- Enhancer Solution
- Wash Buffer (10X)
- phospho-GSK3b (Ser9) Capture Antibody Reagent
- phospho-GSK3b (Ser9) Detection Antibody Reagent
- Positive Control Cell Lysate (lyophilized)
- Detection Reagent (TMB Substrate)
- Stop Solution
- Plate Seal
The InstantOne™ ELISA is specifically engineered for accurate measurement of phosphorylated human and mouse GSK3b (Ser9) in cell lysates. Detection of phosphorylated rat GSK3b (Ser9) is also expected. The InstantOne™ ELISA kit allows for fast analysis of samples in approximately one hour. All reagents used in a traditional sandwich ELISA are added in solution to a plate followed by a wash step and detection with the TMB colorimetric substrate.
Glycogen synthase kinase-3 beta (GSK3 beta) belongs to the subfamily of glycogen synthase kinases, that play a central role in energy metabolism, neuronal cell development, and body pattern formation. GSK3 has alpha and beta isoforms (51 kDa and 47 kDa respectively), that are functionally dissimilar. GSK3 beta plays a role in insulin and Wnt signaling, and is inhibited following the extracellular signaling events.
Tekniske data
Tekniske data
| Adgangsnummer | P49841,Q9WV60 |
| Konjugeret | Biotin |
| Produkttype | ELISA Kit |
| Prøvetype | Cell Lysates |
| Målarter | Human, Mouse |
| Til brug med (applikation) | ELISA |
| Til brug med (udstyr) | Colorimetric Microplate Reader |
| Gen-id (Entrez) | 2932, 56637 |
| Gen symbol | Gsk3b |
| Sættets indhold | Capture Antibody, Detection Antibody Reagent, InstantOne™ Assay Plate, Cell Lysis Buffer, Enhancer Solution, Wash Buffer (10X), Substrate Reagent (TMB), Stop Solution, Positive Control Cell Lysate (lyophilized), Plate seal |
| Vis mere |
For Research Use Only
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion