Få mere at vide
Invitrogen™ Phospho-4EBP1 (Thr37, Thr46) Recombinant Rabbit Monoclonal Antibody (4EB1T37T46-A5)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA527999
 Se flere versioner af dette produkt
Produktkode 15927204
  • 4355.00 DKK / 200µL
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
This gene encodes one member of a family of translation repressor proteins. The protein directly interacts with eukaryotic translation initiation factor 4E, which is a limiting component of the multisubunit complex that recruits 40S ribosomal subunits to the 5' end of mRNAs. Interaction of this protein with eIF4E inhibits complex assembly and represses translation. This protein is phosphorylated in response to various signals including UV irradiation and insulin signaling, resulting in its dissociation from eIF4E and activation of mRNA translation.
TRUSTED_SUSTAINABILITY
Tekniske data
Tekniske data
Phospho-4EBP1 (Thr37, Thr46)
Recombinant Monoclonal
0.5 mg/mL
PBS with 0.1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Q13541, Q60876, Q62622
EIF4EBP1
A synthetic phospho-peptide corresponding to residues surrounding Thr37/46 of human phospho 4E-BP1.
200 μL
Primary
Human, Mouse, Rat
Antibody
IgG κ
Flow Cytometry, Immunohistochemistry, Western Blot
4EB1T37T46-A5
Unconjugated
EIF4EBP1
4EBP1; 4E-BP1; AA959816; BP-1; eIF4E-binding protein 1; EIF4EBP1; eukaryotic translation initiation factor 4E binding protein 1; eukaryotic translation initiation factor 4E-binding protein 1; MGC431; MGC4316; P/OKCL.6; PHAS-1; PHAS-I; Phosphorylated heat- and acid-stable protein regulated by insulin 1
Rabbit
Protein A/G
RUO
116636, 13685, 1978
-20°C
Liquid
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Promotions
Promotions