Få mere at vide
MedChemExpress Phosphatase Inhibitor Cocktail II
Tekniske data
Tekniske data
| Produkttype | Kits |
| Indhold og opbevaring | The product should be stored at -20°C, and is stable for up to 12 months. When kept at 2°C to 8°C, the product remains stable for 2 months. |
| Forsendelsestilstand | Room temperature |
| Mængde | 1 mL |
| Mål | VD/VDR |