Få mere at vide
MedChemExpress Phosphatase Inhibitor Cocktail II
Artikelnummer. 16432479
Encompass_Preferred
Skift visning
:
1 mL
Artikelnummer.
16432479 1 mL
1 options
Artikelnummer. 16432479 Leverandør MedChemExpress Leverandørnr. HYK0022
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
MedChemExpress Phosphatase Inhibitor Cocktail II effectively inhibits acid and alkaline phosphatases as well as protein tyrosine phosphatases(PTPs).
Tekniske data
Produkttype Kits
Indhold og opbevaring The product should be stored at -20°C, and is stable for up to 12 months. When kept at 2°C to 8°C, the product remains stable for 2 months.
Forsendelsestilstand Room temperature
Mængde 1 mL
Mål VD/VDR