Få mere at vide
Phosmet-d6, TRC
Artikelnummer. 30240837
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30240837 10 mg
30240829 50 mg
30240557 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30240837 Leverandør Toronto Research Chemicals Leverandørnr. TRCP35370210MG
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2083623-41-8
Molekylær formel C11 2H6 H6 N O4 P S2
Molekylvægt (g/mol) 323.36
Synonym Phosmet D6 (dimethyl D6), Phosphorodithioic acid, S-[(1,3-dihydro-1,3-dioxo-2H-isoindol-2-yl)methyl] O,O-di(methyl-d3) ester, Phosmet D6 (dimethoxy D6)
IUPAC navn 2-[bis(trideuteriomethoxy)phosphinothioylsulfanylmethyl]isoindole-1,3-dione
SMIL [2H]C([2H])([2H])OP(=S)(OC([2H])([2H])[2H])SCN1C(=O)c2ccccc2C1=O
Tekniske data
Kemisk navn eller materiale Phosmet D6 (dimethoxy D3)
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C11H12NO4PS2/c1-15-17(18,16-2)19-7-12-10(13)8-5-3-4-6-9(8)11(12)14/h3-6H,7H2,1-2H3/i1D3,2D3
Formel vægt 323.0322
Procent renhed >95%