Få mere at vide
Phosacetim, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Phosacetim |
| CAS | 4104-14-7 |
| Molekylær formel | C14H13Cl2N2O2PS |
| Mængde | 2.5 g |
| Synonym | N-(1-Iminoethyl)-phosphoramidothioic Acid O,O-Bis(4-chlorophenyl) Ester, (1-Iminoethyl)-phosphoramidothioic Acid O,O-Bis(4-chlorophenyl) Ester, Acetimidoyl-phosphoramidothioic Acid O,O-Bis(p-chlorophenyl) Ester, BAY 38819, Bayer 38819, DRC 714, Gophacide, O,O-Bis(p-chlorophenyl) Acetimidoylamidothiophosphate, O,O-Bis(p-chlorophenyl) Acetimidoylphosphoramidothioate |
| InChI formel | InChI=1S/C14H13Cl2N2O2PS/c1-10(17)18-21(22,19-13-6-2-11(15)3-7-13)20-14-8-4-12(16)5-9-14/h2-9H,1H3,(H2,17,18,22) |
| SMIL | CC(=N)NP(=S)(Oc1ccc(Cl)cc1)Oc2ccc(Cl)cc2 |
| IUPAC navn | N-bis(4-chlorophenoxy)phosphinothioylethanimidamide |
| Molekylvægt (g/mol) | 375.21 |
| Formel vægt | 373.9812 |