Få mere at vide
Phorate 1000 μg/mL in Acetone, Dr. Ehrenstorfer
Artikelnummer. 30308936
Skift visning
Mængde:
1 ml
Pakningsstørrelse:
1ml
Artikelnummer. Mængde unitSize
30308936 1 ml 1ml
1 options
Artikelnummer. 30308936 Leverandør Dr. Ehrenstorfer Leverandørnr. DREA16080000AC1000
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phorate
CAS 298-02-2
Analyt- eller komponentnavne Phorate
Forsendelsestilstand +20°C
Molekylær formel C7 H17 O2 P S3
Koncentration 1000 μg/mL
Certificeringer/compliance ISO/IEC 17025
Synonym Phosphorodithioic acid, O,O-diethyl S-[(ethylthio)methyl] ester, American Cyanamid 3911, ENT 24,042, Experimental Insecticide 3911, Granutox, L 11/6, O,O-Diethyl S-[(ethylthio)methyl] phosphorodithioate, O,O-Diethyl S-ethylmercaptomethyl dithiophosphate, O,O-Diethyl S-ethylthiomethyl dithiophosphate, Phorate, Phorate 10G, Thimet, Thimet 10G, Thimet G, Timet, VUAgT 182, suScon FuMing
InChI formel InChI=1S/C7H17O2PS3/c1-4-8-10(11,9-5-2)13-7-12-6-3/h4-7H2,1-3H3
Løsningstype Single Solution
SMIL CCOP(=S)(OCC)SCSCC
IUPAC navn diethoxy-(ethylsulfanylmethylsulfanyl)-sulfanylidene-λ^{5}-phosphane
Opløsningsmiddel eller Matrix Acetone
Industry Type Environmental Testing
Molekylvægt (g/mol) 260.38
Mængde 1 ml
Formel vægt 260.0128 g/mol