Få mere at vide
Thermo Scientific™ Phire Plant Direct PCR Master Mix PROMO
Amplify DNA directly from plant samples without prior DNA purification with Phire Plant Direct PCR Master Mix
Brand: Thermo Scientific™ F160S-RMD123
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Highlights- Direct PCR - sample is added directly to PCR reaction, therefore there is no need for time-consuming and expensive DNA purification steps.
- Dilution & Storage protocol available - allows tens of PCR reactions from one tiny sample and allows re-testing
- Specially engineered Phire Hot Start II DNA polymerase - extremely short PCR protocol times
- Master Mix format with premixed gel loading dye - minimizes possibility of cross-contamination, reduces sample handling and allows direct loading on gel
- Genotyping
- Transgene detection
- Knockout analysis
- Sequencing
- 2x Phire Plant Direct PCR Master Mix
- Dilution Buffer
- Control Primer Mix (25 µM each)
- Water, nuclease- free
- O'GeneRuler Express DNA Ladder