Få mere at vide
Thermo Scientific™ Phire Plant Direct PCR Master Mix PROMO
Amplify DNA directly from plant samples without prior DNA purification with Phire Plant Direct PCR Master Mix
Brand:  Thermo Scientific™ F160S-RMD123
Produktkode 15619464
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
Highlights
  • Direct PCR - sample is added directly to PCR reaction, therefore there is no need for time-consuming and expensive DNA purification steps.
  • Dilution & Storage protocol available - allows tens of PCR reactions from one tiny sample and allows re-testing
  • Specially engineered Phire Hot Start II DNA polymerase - extremely short PCR protocol times
  • Master Mix format with premixed gel loading dye - minimizes possibility of cross-contamination, reduces sample handling and allows direct loading on gel
Applications
  • Genotyping
  • Transgene detection
  • Knockout analysis
  • Sequencing
Includes
  • 2x Phire Plant Direct PCR Master Mix
  • Dilution Buffer
  • Control Primer Mix (25 µM each)
  • Water, nuclease- free
  • O'GeneRuler Express DNA Ladder
TRUSTED_SUSTAINABILITY
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner