Få mere at vide
Uvex™ Pheos B-WR Safety Helmet
Shop alle Uvex produkter
Farve:
Black
White
Beskrivelse
- Resistant to hot molten metal splash and temperatures down to -30°C (EN 367 additional requirement MM)
- Side euroslot adapters (30 mm) are suitable for the addition of earmuffs or the uvex pheos visor system
- Suspension harness with wheel ratchet allows continuous width adjustment, for a perfect fit at all times
- Six-point textile band attachment ensures optimal fit and comfort
- Available in a wide variety of color options
Tekniske data
Tekniske data
| Farve | Black |
| Model- | Pheos B-WR |
| Ventileret | Ventilated |