Få mere at vide
Phenyramidol Salicylate, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Phenyramidol Salicylate |
| CAS | 1235-07-0 |
| Analyt- eller komponentnavne | Phenyramidol Salicylate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C13 H14 N2 O . C7 H6 O3 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Analexin syrup, Phenyramidol salicylate, Benzoic acid, 2-hydroxy-, compd. with alpha-[(2-pyridinylamino)methyl]benzenemethanol (1:1), Salicylic acid, compd. with alpha-[(2-pyridylamino)methyl]benzyl alcohol (1:1) (8CI), Salicylic acid, compd. with alpha-[(2-pyridylamino)methyl]benzyl alcohol (7CI), Benzenemethanol, alpha-[(2-pyridinylamino)methyl]-, mono(2-hydroxybenzoate) (salt) (9CI), Benzyl alcohol, alpha-[(2-pyridylamino)methyl]-, monosalicylate (salt) |
| InChI formel | InChI=1S/C13H14N2O.C7H6O3/c16-12(11-6-2-1-3-7-11)10-15-13-8-4-5-9-14-13;8-6-4-2-1-3-5(6)7(9)10/h1-9,12,16H,10H2,(H,14,15);1-4,8H,(H,9,10) |
| Løsningstype | Neat |
| SMIL | OC(CNc1ccccn1)c2ccccc2.OC(=O)c3ccccc3O |
| Vis mere |