missing translation for 'onlineSavingsMsg'
Learn More

Phenyltrichlorosilane, 95%, Thermo Scientific Chemicals

Catalog Number p-184210
Click to view available options
Quantity:
10 mL
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
98-13-5
C6H5Cl3Si
211.55
MFCD00000479
ORVMIVQULIKXCP-UHFFFAOYSA-N
trichloro phenyl silane, phenyltrichlorosilane, silane, trichlorophenyl, phenyl trichlorosilane, phenylsilicon trichloride, silane, phenyltrichloro, silicon phenyl trichloride, benzene, trichlorosilyl, unii-m7199jl06m, phenyl-trichloro-silane
7372
trichloro(phenyl)silane
C1=CC=C(C=C1)[Si](Cl)(Cl)Cl
This item is not returnable. View return policy
98-13-5
C6H5Cl3Si
211.55
MFCD00000479
ORVMIVQULIKXCP-UHFFFAOYSA-N
trichloro phenyl silane, phenyltrichlorosilane, silane, trichlorophenyl, phenyl trichlorosilane, phenylsilicon trichloride, silane, phenyltrichloro, silicon phenyl trichloride, benzene, trichlorosilyl, unii-m7199jl06m, phenyl-trichloro-silane
7372
trichloro(phenyl)silane
C1=CC=C(C=C1)[Si](Cl)(Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.3210g/mL
Boiling Point 201°C
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula C6H5SiCl3
Refractive Index 1.5220 to 1.524
Quantity 10 mL
Beilstein 16, 911
Specific Gravity 1.321
Solubility Information Solubility in water: reacts. Other solubilities: soluble in benzene,ehter and perchloroethylene
Formula Weight 211.55
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Phenyltrichlorosilane, 95%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Phenyltrichlorosilane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • EUH014-Reacts violently with water.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P232-Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'