Få mere at vide
Phenyltoloxamine Citrate, Mikromol™
Artikelnummer. 30049524
Skift visning
Mængde:
500 mg
Artikelnummer. Mængde
30049524 500 mg
1 options
Artikelnummer. 30049524 Leverandør Mikromol Leverandørnr. MM0770.00
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Phenyltoloxamine Citrate
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phenyltoloxamine Citrate
CAS 1176-08-5
Analyt- eller komponentnavne Phenyltoloxamine Citrate
Forsendelsestilstand Room Temperature
Molekylær formel C17 H21 N O . C6 H8 O7
Certificeringer/compliance ISO 17034
Synonym Phenyltoloxamine Citrate
InChI formel InChI=1S/C17H21NO.C6H8O7/c1-18(2)12-13-19-17-11-7-6-10-16(17)14-15-8-4-3-5-9-15;7-3(8)1-6(13,5(11)12)2-4(9)10/h3-11H,12-14H2,1-2H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)
SMIL CN(C)CCOc1ccccc1Cc2ccccc2.OC(=O)CC(O)(CC(=O)O)C(=O)O
IUPAC navn 2-(2-benzylphenoxy)-N,N-dimethylethanamine;2-hydroxypropane-1,2,3-tricarboxylic acid
Molekylvægt (g/mol) 447.48
Mængde 500 mg
Formel vægt 447.1893 g/mol