missing translation for 'onlineSavingsMsg'
Learn More

Phenylphosphonic dichloride, 97%, Thermo Scientific Chemicals

Catalog Number p-183663
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
824-72-6
C6H5Cl2OP
194.98
MFCD00002070
IBDMRHDXAQZJAP-UHFFFAOYSA-N
phenylphosphonic dichloride, phosphonic dichloride, phenyl, benzenephosphonic dichloride, phenylphosphonyl dichloride, dichlorophenylphosphine oxide, phenyldichlorophosphine oxide, benzenephosphonyl chloride, phenylphosphonic acid dichloride, phenylphosphonodichloridic acid, phosphonic dichloride, p-phenyl
69990
dichlorophosphorylbenzene
ClP(Cl)(=O)C1=CC=CC=C1
This item is not returnable. View return policy
824-72-6
C6H5Cl2OP
194.98
MFCD00002070
IBDMRHDXAQZJAP-UHFFFAOYSA-N
phenylphosphonic dichloride, phosphonic dichloride, phenyl, benzenephosphonic dichloride, phenylphosphonyl dichloride, dichlorophenylphosphine oxide, phenyldichlorophosphine oxide, benzenephosphonyl chloride, phenylphosphonic acid dichloride, phenylphosphonodichloridic acid, phosphonic dichloride, p-phenyl
69990
dichlorophosphorylbenzene
ClP(Cl)(=O)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 3.0°C
Color Yellow to Brown
Density 1.3900g/mL
Boiling Point 258.0°C
Flash Point 204°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Linear Formula C6H5P(O)Cl2
Refractive Index 1.5570 to 1.5610
Quantity 250 mL
Beilstein 16, 804
Fieser 01,846; 16,284
Specific Gravity 1.39
Solubility Information Solubility in water: reacts
Formula Weight 194.98
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Phenylphosphonic dichloride, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'