Få mere at vide
Phenylmethyl Ester 2-[1-(3,4-Dimethoxybenzoyl)-2-(1-ethylpropoxy)-1-methyl-2-oxoethyl] Hydrazinecarboxylic Acid, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Phenylmethyl Ester 2-[1-(3,4-Dimethoxybenzoyl)-2-(1-ethylpropoxy)-1-methyl-2-oxoethyl] Hydrazinecarboxylic Acid |
| CAS | 1281895-71-3 |
| Molekylær formel | C25H32N2O7 |
| Mængde | 10 mg |
| Synonym | 2-[1-(3,4-Dimethoxybenzoyl)-2-(1-ethylpropoxy)-1-methyl-2-oxoethyl]-phenylmethyl Ester Hydrazinecarboxylic acidPhenylmethyl 2-[1-(3,4-Dimethoxybenzoyl)-2-(1-ethylpropoxy)-1-methyl-2-oxoethyl]hydrazinecarboxylate |
| InChI formel | InChI=1S/C25H32N2O7/c1-6-19(7-2)34-23(29)25(3,22(28)18-13-14-20(31-4)21(15-18)32-5)27-26-24(30)33-16-17-11-9-8-10-12-17/h8-15,19,27H,6-7,16H2,1-5H3,(H,26,30)/t25-/m1/s1 |
| SMIL | CCC(CC)OC(=O)[C@](C)(NNC(=O)OCc1ccccc1)C(=O)c2ccc(OC)c(OC)c2 |
| IUPAC navn | pentan-3-yl (2R)-3-(3,4-dimethoxyphenyl)-2-methyl-3-oxo-2-(2-phenylmethoxycarbonylhydrazinyl)propanoate |
| Molekylvægt (g/mol) | 472.54 |
| Formel vægt | 472.221 |