Få mere at vide
Phenylglyoxylic Acid-d5, TRC
Artikelnummer. 30409013
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30409013 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30409013 Leverandør Toronto Research Chemicals Leverandørnr. TRCP3275025MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phenylglyoxylic Acid-d5
CAS 1217089-53-6
Renhedsgrad noter HPLC
Molekylær formel C8HD5O3
Mængde 5 mg
Synonym α-Oxobenzeneacetic Acid-d5, 2-Oxo-2-(phenyl-d5)acetic Acid, 2-Oxo-2-(phenyl-d5)ethanoic Acid, Benzoylformic Acid-d5, α-Ketophenylacetic Acid-d5, NSC 28293-d5
InChI formel InChI=1S/C8H6O3/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5H,(H,10,11)/i1D,2D,3D,4D,5D
SMIL OC(C(C1=C([2H])C([2H])=C([2H])C([2H])=C1[2H])=O)=O
Molekylvægt (g/mol) 155.16
Formel vægt 155.0631
Procent renhed >95