Få mere at vide
Phenylglycine Ethyl Ester Hydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Phenylglycine Ethyl Ester Hydrochloride |
| CAS | 879-48-1 |
| Molekylær formel | C10H14ClNO2 |
| Mængde | 25 mg |
| Synonym | Ethyl Phenylglycinate Hydrochloride, DL-Phenylglycine Ethyl Ester Hydrochloride, alpha-Amino-benzeneacetic Acid Ethyl Ester Hydrochloride, NSC 24595 |
| InChI formel | InChI=1S/C10H13NO2.ClH/c1-2-13-10(12)9(11)8-6-4-3-5-7-8;/h3-7,9H,2,11H2,1H3;1H |
| SMIL | Cl.CCOC(=O)C(N)c1ccccc1 |
| IUPAC navn | ethyl 2-amino-2-phenylacetate;hydrochloride |
| Molekylvægt (g/mol) | 215.68 |
| Formel vægt | 215.0713 |