Få mere at vide
Phenylephrone-d3 Hydrochloride, TRC
Artikelnummer. 30422674
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30422674 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30422674 Leverandør Toronto Research Chemicals Leverandørnr. TRCP32067210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phenylephrone-d3 Hydrochloride
CAS 1246819-29-3
Molekylær formel C9H9D3ClNO2
Mængde 10 mg
Synonym 1-(3-Hydroxyphenyl)-2-(methyl-d3-amino)ethanone Hydrochloride, 3'-Hydroxy-2-(methyl-d3)aminoacetophenone Hydrochloride, Phenylephrine Imp. C (EP)-d3','Hydrochloride
InChI formel InChI=1S/C9H11NO2.ClH/c1-10-6-9(12)7-3-2-4-8(11)5-7;/h2-5,10-11H,6H2,1H3;1H/i1D3;
SMIL OC1=CC=CC(C(CNC([2H])([2H])[2H])=O)=C1.Cl
Molekylvægt (g/mol) 204.67
Formel vægt 204.0745