Få mere at vide
Phenylbutazone EP Impurity A, Mikromol™
Artikelnummer. 30051154
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051154 25 mg
30050208 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051154 Leverandør Mikromol Leverandørnr. MM0548.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Phenylbutazone EP Impurity A
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2RS)-2-[(1,2-Diphenyldiazanyl)carbonyl]hexanoic Acid
CAS 3583-64-0
Analyt- eller komponentnavne (2RS)-2-[(1,2-Diphenyldiazanyl)carbonyl]hexanoic Acid
Forsendelsestilstand Ice pack (-20°C)
Molekylær formel C19 H22 N2 O3
Certificeringer/compliance ISO 17025
Synonym (2RS)-2-[(1,2-Diphenyldiazanyl)carbonyl]hexanoic Acid, Phenylbutazone Imp. A (EP), Propanedioic acid, 2-butyl-, 1-(1,2-diphenylhydrazide), Malonic acid, butyl-, 1,2-diphenylhydrazide (7CI), Malonic acid, butyl-, mono(1,2-diphenylhydrazide) (8CI), Propanedioic acid, butyl-, mono(1,2-diphenylhydrazide) (9CI), Bumadizon, Bumadizone, Butylmalonic acid mono(1,2-diphenylhydrazide), N-(2-Carboxycaproyl)hydrazobenzene, Phenylbutazone Impurity A
InChI formel InChI=1S/C19H22N2O3/c1-2-3-14-17(19(23)24)18(22)21(16-12-8-5-9-13-16)20-15-10-6-4-7-11-15/h4-13,17,20H,2-3,14H2,1H3,(H,23,24)
SMIL CCCCC(C(=O)O)C(=O)N(Nc1ccccc1)c2ccccc2
IUPAC navn 2-[anilino(phenyl)carbamoyl]hexanoic acid
Molekylvægt (g/mol) 326.39
Mængde 25 mg
Formel vægt 326.163 g/mol