missing translation for 'onlineSavingsMsg'
Få mere at vide

Phenylboronic Acid-d5, TRC

Artikelnummer. 30414940
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30414940 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30414940 Leverandør Toronto Research Chemicals Leverandørnr. TRCP3195921G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Phenylboronic Acid-d5
CAS 215527-70-1
Molekylær formel C6H2D5BO2
Mængde 1 g
Synonym B-Phenylboronic Acid-d5, Dihydroxyphenylborane-d5, NSC 66487-d5, Phenylboric Acid-d5, Phenylboronic Acid-d5, Phenyldihydroxyborane-d5, B-(Phenyl-2,3,4,5,6-d5)boronic Acid, Phenyl-d5-boronic Acid, (Pentadeuterophenyl)boronic Acid, (Perdeuterophenyl)boronic Acid
InChI formel InChI=1S/C6H7BO2/c8-7(9)6-4-2-1-3-5-6/h1-5,8-9H/i1D,2D,3D,4D,5D
SMIL OB(C1=C([2H])C([2H])=C([2H])C([2H])=C1[2H])O
Molekylvægt (g/mol) 126.96
Formel vægt 127.0853
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.