Få mere at vide
Phenylbis(2,4,6-trimethylbenzoyl)phosphine Oxide, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Phenylbis(2,4,6-trimethylbenzoyl)phosphine Oxide |
| CAS | 162881-26-7 |
| Molekylær formel | C26H27O3P |
| Mængde | 1 g |
| Synonym | 1,1'-(Phenylphosphinylidene)bis[1-(2,4,6-trimethylphenyl)methanone] (ACI), Phosphine oxide','phenylbis(2,4,6-trimethylbenzoyl)- (9CI), (Phenylphosphoryl)bis(mesitylmethanone), 1,1'-(Phenylphosphinylidene)bis[(2,4,6-trimethylphenyl)methanone], 819DW, [Phenyl-(2,4,6-trimethylbenzoyl)phosphoryl]-(2,4,6-trimethylphenyl)methanone, BAPO, BAPO 1, BASF 819, Bis(2,4,6-trimethylbenzoyl)phenylphosphine oxide, Boxing 819, CGI 819XF, Chivacure 789, Ciba 819, Daido UV-Cure 819, Darocur 819, Doublecure 1819, Genocure BAPO, GR-XBPO, I 819, IC 819, IGM 819, IR 819, IRG 819, Irgacure 801, Irgacure 819, Irgacure 819DW, Irgacure 919, Irgacure 981, Irgacure I 819, JRCure 1109, JRCure 1819, JRCure 819, Keycure 981, Lunacure 900, Omnicure 819, Omnirad 380, Omnirad 819, Omnirad 819DW, Omnirad 851, Omnirad 981, P 2312, Phenylbis(2,4,6-trimethylbenzoyl)phosphine oxide, Phenylbis(2,4,6-trimethylbenzyl)phosphine oxide, Photocuring agent 819, Photoinitiator 819, Photoinitiator 850, Photoinitiator 898, PI 819, Ryoji 819, SB-PI 719, Speedcure 819, Speedcure BPO, UV-Cure 819, XBPO |
| InChI formel | InChI=1S/C26H27O3P/c1-16-12-18(3)23(19(4)13-16)25(27)30(29,22-10-8-7-9-11-22)26(28)24-20(5)14-17(2)15-21(24)6/h7-15H,1-6H3 |
| SMIL | Cc1cc(C)c(C(=O)P(=O)(C(=O)c2c(C)cc(C)cc2C)c3ccccc3)c(C)c1 |
| IUPAC navn | [phenyl-(2,4,6-trimethylbenzoyl)phosphoryl]-(2,4,6-trimethylphenyl)methanone |
| Molekylvægt (g/mol) | 418.46 |
| Formel vægt | 418.1698 |