Få mere at vide
Phenylacetyl-d5 L-Glutamine, TRC
Artikelnummer. 30424333
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30424333 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30424333 Leverandør Toronto Research Chemicals Leverandørnr. TRCP30650210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phenyl-d5-acetyl L-Glutamine
CAS 1331909-01-3
Renhedsgrad noter HPLC
Molekylær formel C13 D5 H11 N2 O4
Mængde 10 mg
Synonym L-Glutamine','N2-[2-(phenyl-2,3,4,5,6-d5)acetyl]- (ACI), N2-[2-(Phenyl-2,3,4,5,6-d5)acetyl]-L-glutamine (ACI), (2S)-5-Amino-5-oxo-2-[[2-(2,3,4,5,6-pentadeuteriophenyl)acetyl]amino]pentanoic acid, Nα-(Phenyl-d5-acetyl)-L-glutamine, Phenylacetyl-d5 L-Glutamine, (2S)-5-Amino-5-oxo-2-[2-(phenyl-d5)acetamido]pentanoic acid, N-Phenylacetyl-L-glutamine-d5, N-Phenylacetyl-L-glutamine-D5 (phenyl-D5)
InChI formel InChI=1S/C13H16N2O4/c14-11(16)7-6-10(13(18)19)15-12(17)8-9-4-2-1-3-5-9/h1-5,10H,6-8H2,(H2,14,16)(H,15,17)(H,18,19)/t10-/m0/s1/i1D,2D,3D,4D,5D
SMIL [2H]c1c([2H])c([2H])c(CC(=O)N[C@@H](CCC(=O)N)C(=O)O)c([2H])c1[2H]
IUPAC navn (2S)-5-amino-5-oxo-2-[[2-(2,3,4,5,6-pentadeuteriophenyl)acetyl]amino]pentanoic acid
Molekylvægt (g/mol) 269.308
Formel vægt 269.142
Procent renhed >95