Få mere at vide
Phenylacetyl-d5 Chloride, TRC
Artikelnummer. 30335438
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30335438 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30335438 Leverandør Toronto Research Chemicals Leverandørnr. TRCB188802100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phenylacetyl-d5 Chloride
CAS 216966-11-9
Molekylær formel C8H2D5ClO
Mængde 100 mg
Synonym 2-Phenylacetyl-d5 Chloride, 2-Phenylethanoyl-d5 Chloride, Phenylacetic-d5 Acid Chloride, Phenylacetic-d5 Chloride, Phenylacetyl-d5 Chloride, α-Phenylacetyl-d5 Chloride
InChI formel InChI=1S/C8H7ClO/c9-8(10)6-7-4-2-1-3-5-7/h1-5H,6H2/i1D,2D,3D,4D,5D
SMIL ClC(CC1=C([2H])C([2H])=C([2H])C([2H])=C1[2H])=O
IUPAC navn 2-(2,3,4,5,6-pentadeuteriophenyl)acetyl chloride
Molekylvægt (g/mol) 159.62