Få mere at vide
Phenyl Tosylate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Phenyl Tosylate |
| CAS | 640-60-8 |
| Molekylær formel | C13 H12 O3 S |
| Mængde | 1 g |
| Synonym | p-Toluenesulfonic Acid Phenyl Ester, NSC 10407, NSC 14917, Phenyl 4-Methylbenzenesulfonate, Phenyl 4-Toluenesulfonate, Phenyl p-Methylbenzenesulfonate, Phenyl p-Toluenesulfonate, Phenyl Toluenesulfonate, Phenyl Tosylate |
| InChI formel | InChI=1S/C13H12O3S/c1-11-7-9-13(10-8-11)17(14,15)16-12-5-3-2-4-6-12/h2-10H,1H3 |
| SMIL | Cc1ccc(cc1)S(=O)(=O)Oc2ccccc2 |
| IUPAC navn | phenyl 4-methylbenzenesulfonate |
| Molekylvægt (g/mol) | 248.298 |
| Formel vægt | 248.051 |