Få mere at vide
(Phenyl sulfone)-d10, TRC
Artikelnummer. 30414908
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30414908 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30414908 Leverandør Toronto Research Chemicals Leverandørnr. TRCP218851100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (Phenyl sulfone)-d10
CAS 21885-27-8
Molekylær formel C12 D10 O2 S
Mængde 100 mg
InChI formel InChI=1S/C12H10O2S/c13-15(14,11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D
SMIL [2H]c1c([2H])c([2H])c(c([2H])c1[2H])S(=O)(=O)c2c([2H])c([2H])c([2H])c([2H])c2[2H]
IUPAC navn 1,2,3,4,5-pentadeuterio-6-(2,3,4,5,6-pentadeuteriophenyl)sulfonylbenzene
Molekylvægt (g/mol) 228.333
Formel vægt 228.103